Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC mAChR-IN-1 hydrochloride | 119391-73-0 | MFCD01725710 | 99.6% | 524.82 g/mol | C23H26ClIN2O2 | 10MG
SDP

Catalog No. 5000237894
Encompass_Preferred

mAChR-IN-1 hydrochloride is a research-grade muscarinic acetylcholine receptor antagonist provided as the hydrochloride salt. It exhibits high potency (IC50 = 17 nM) and is characterized by a defined molecular formula and high reported purity, making it suitable for in vitro pharmacology and biochemical assays.

  • Potent mAChR antagonist (IC50 = 17 nM).
  • Hydrochloride salt form for consistent handling.
  • High reported purity (99.6%).
  • Molecular weight 524.82 g/mol; formula C23H26ClIN2O2.
  • Intended for in vitro research applications.

Catalog No. 50-002-37894 Supplier Medchemexpress LLC Supplier No. HY12426A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

mAChR-IN-1 hydrochloride is a research-grade muscarinic acetylcholine receptor antagonist provided as the hydrochloride salt. It exhibits high potency (IC50 = 17 nM) and is characterized by a defined molecular formula and high reported purity, making it suitable for in vitro pharmacology and biochemical assays.

  • Potent mAChR antagonist (IC50 = 17 nM).
  • Hydrochloride salt form for consistent handling.
  • High reported purity (99.6%).
  • Molecular weight 524.82 g/mol; formula C23H26ClIN2O2.
  • Intended for in vitro research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.