Learn More
Medchemexpress LLC Macimorelin (acetate) | 945212-59-9 | 99.3% | 534.61 | 5 MG
SDP
Catalog No. 5000363659
Encompass_Preferred
Macimorelin (EP-1572) acetate is an orally active GHSR agonist and a GH secretagogue that stimulates GH release. It is utilized in research for adult growth hormone deficiency (AGHD) and Cancer anorexia-cachexia syndrome (CACS).
  • Orally active GHSR agonist
  • Stimulates GH release
  • Used in adult growth hormone deficiency (AGHD) research
  • Used in cancer anorexia-cachexia syndrome (CACS) research
Catalog No. 50-003-63659 Supplier Medchemexpress LLC Supplier No. HY14820A5MG
Macimorelin (EP-1572) acetate is an orally active GHSR agonist and a GH secretagogue that stimulates GH release. It is utilized in research for adult growth hormone deficiency (AGHD) and Cancer anorexia-cachexia syndrome (CACS).
  • Orally active GHSR agonist
  • Stimulates GH release
  • Used in adult growth hormone deficiency (AGHD) research
  • Used in cancer anorexia-cachexia syndrome (CACS) research