Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Macimorelin acetate | 945212-59-9 | MFCD09260160 | 99.3% | 534.61 g·mol⁻¹ | C28H34N6O5 | 10 MG
SDP

Catalog No. 5000286017
Encompass_Preferred

Macimorelin acetate is an orally active ghrelin receptor (GHSR) agonist and growth hormone secretagogue used in research to stimulate growth hormone release. It is applied in studies of adult growth hormone deficiency and cancer anorexia-cachexia syndrome.

  • Orally active GHSR agonist that stimulates growth hormone release.
  • Used in research on adult growth hormone deficiency and cancer anorexia-cachexia syndrome.
  • High reported purity (99.34%).
  • Molecular formula C28H34N6O5 and molecular weight 534.61 g·mol⁻¹.
  • Available in mg-scale quantities suitable for laboratory research.
  • Product documentation available: datasheet, COA, and SDS for handling and safety.

Catalog No. 50-002-86017 Supplier Medchemexpress LLC Supplier No. HY14820A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Macimorelin acetate is an orally active ghrelin receptor (GHSR) agonist and growth hormone secretagogue used in research to stimulate growth hormone release. It is applied in studies of adult growth hormone deficiency and cancer anorexia-cachexia syndrome.

  • Orally active GHSR agonist that stimulates growth hormone release.
  • Used in research on adult growth hormone deficiency and cancer anorexia-cachexia syndrome.
  • High reported purity (99.34%).
  • Molecular formula C28H34N6O5 and molecular weight 534.61 g·mol⁻¹.
  • Available in mg-scale quantities suitable for laboratory research.
  • Product documentation available: datasheet, COA, and SDS for handling and safety.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.