Learn More
Cayman Chemical MacImorelInacetate 50mg
SDP
Catalog No. 502943868
Encompass_Preferred
A growth hormone secretagogue and ghrelin receptor agonist; binds to GHS-R1a (IC50 = 123 nM for the human receptor); oral administration increases plasma growth hormone levels in dogs at 1 mg/kg; decreases gastric acid secretion in pylorus-ligated rats at 0.01 or 1 nmol/animal, i.c.v.; increases acute food intake and body weight in mice at 10 mg/kg; decreases the number and total duration of seizures in mice induced with dopamine D1 receptor kindling using SKF 81297
Catalog No. 50-294-3868 Supplier Cayman Chemical Supplier No. 4188650 MG
A growth hormone secretagogue and ghrelin receptor agonist; binds to GHS-R1a (IC50 = 123 nM for the human receptor); oral administration increases plasma growth hormone levels in dogs at 1 mg/kg; decreases gastric acid secretion in pylorus-ligated rats at 0.01 or 1 nmol/animal, i.c.v.; increases acute food intake and body weight in mice at 10 mg/kg; decreases the number and total duration of seizures in mice induced with dopamine D1 receptor kindling using SKF 81297