Learn More
Apexbio Technology LLC Macitentan, 25mg. Cas: 441798-33-0 MFCD: MFCD17167076. Endothelin (ET)(A) and ET(B) receptor antagonist
SDP
Supplier:  Apexbio Technology LLC A192925
Encompass_Preferred
Macitentan is a new dual ETA/ETB endothelin (ET) receptor antagonist, with mean IC50 values of 0.5 0.2 nM (n= 17) to inhibit the binding of 125I-ET-1 to recombinant ETA receptors, and of 391182 nM (n= 17) for ETB receptors in Chinese hamster ovary cells [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4725