Learn More
Medchemexpress LLC HY-14184 50mg , Macitentan CAS:441798-33-0 Purity:>98%
SDP
Catalog No. 502259732
Encompass_Preferred
HY-14184 50mg Macitentan CAS: 441798-33-0 Macitentan (ACT-064992) is an orally active, non-peptide dual ETA and ETB (endothelin receptor) antagonist. Macitentan has the potential for idiopathic pulmonary fibrosis (IPF) and pulmonary arterial hypertension (PAH). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9732 Supplier Medchemexpress LLC Supplier No. HY1418450MG
HY-14184 50mg Macitentan CAS: 441798-33-0 Macitentan (ACT-064992) is an orally active, non-peptide dual ETA and ETB (endothelin receptor) antagonist. Macitentan has the potential for idiopathic pulmonary fibrosis (IPF) and pulmonary arterial hypertension (PAH). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.