Learn More
Medchemexpress LLC Macitentan-d4 | 1258428-05-5 | 98.0% | 592.30 | C19H16D4Br2N6O4S | 5 MG
SDP
Catalog No. 5000221353
Encompass_Preferred
Macitentan-d4 is a deuterium-labeled analogue of macitentan provided as an isotope-labeled reference standard for analytical and research applications, particularly as an internal standard for LC-MS or GC-MS quantification and method development.
  • High isotopic labeling for accurate mass spectrometry quantification.
  • Analytical-grade purity suitable for reference standards.
  • Small pack size convenient for assay development and validation.
  • Documented chemical and molecular data for traceability.
  • Stable when stored per recommended conditions.
Catalog No. 50-002-21353 Supplier Medchemexpress LLC Supplier No. HY14184S5MG
Macitentan-d4 is a deuterium-labeled analogue of macitentan provided as an isotope-labeled reference standard for analytical and research applications, particularly as an internal standard for LC-MS or GC-MS quantification and method development.
  • High isotopic labeling for accurate mass spectrometry quantification.
  • Analytical-grade purity suitable for reference standards.
  • Small pack size convenient for assay development and validation.
  • Documented chemical and molecular data for traceability.
  • Stable when stored per recommended conditions.