Learn More
Medchemexpress LLC HY-14184A 5mg Medchemexpress, Macitentan (n-butyl analogue) CAS:556797-16-1 Purity:>98%
SDP
Catalog No. 501932698
Encompass_Preferred
Medchemexpress, HY-14184A 5mg Macitentan (n-butyl analogue) CAS:556797-16-1 Macitentan n-butyl analogue is a n-butyl analogue of Macitentan. Macitentan is an orally active, non-peptide dual endothelin ETA and ETB receptor antagonist for the potential treatment of idiopathic pulmonary fibrosis (IPF) and pulmonary arterial hypertension (PAH). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2698 Supplier Medchemexpress LLC Supplier No. HY14184A5MG
Medchemexpress, HY-14184A 5mg Macitentan (n-butyl analogue) CAS:556797-16-1 Macitentan n-butyl analogue is a n-butyl analogue of Macitentan. Macitentan is an orally active, non-peptide dual endothelin ETA and ETB receptor antagonist for the potential treatment of idiopathic pulmonary fibrosis (IPF) and pulmonary arterial hypertension (PAH). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.