Learn More
Medchemexpress LLC Macitentan (n-butyl analogue) | 556797-16-1 | 99.4% | C20H21Br2N5O4S | 10 MM*ML
SDP
Catalog No. 5000356889
Encompass_Preferred
Macitentan n-butyl analogue is a n-butyl analogue of Macitentan. Macitentan is an orally active, non-peptide dual endothelin ETA and ETB receptor antagonist for the potential treatment of idiopathic pulmonary fibrosis (IPF) and pulmonary arterial hypertension (PAH).
Catalog No. 50-003-56889 Supplier Medchemexpress LLC Supplier No. HY14184A10MM/1ML
Macitentan n-butyl analogue is a n-butyl analogue of Macitentan. Macitentan is an orally active, non-peptide dual endothelin ETA and ETB receptor antagonist for the potential treatment of idiopathic pulmonary fibrosis (IPF) and pulmonary arterial hypertension (PAH).