Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Macitentan (n-butyl analogue) | 556797-16-1 | MFCD28900696 | 99.4% | C20H21Br2N5O4S | 10MG
SDP

Catalog No. 5000260896
Encompass_Preferred

Macitentan (n-butyl analogue) is an n-butyl analogue of macitentan that acts as a dual endothelin ETA and ETB receptor antagonist. It is intended for research use in in vitro and preclinical pharmacology studies focused on endothelin signaling. Not for human or veterinary use.

  • Dual endothelin ETA and ETB receptor antagonist.
  • High purity (99.35%).
  • Molecular formula C20H21Br2N5O4S.
  • Soluble in DMSO; available as 10 mM solutions.
  • Available in small research pack sizes for screening.
  • Suitable for in vitro and preclinical pharmacology studies.
  • Provided as solid and solution formats for flexible use.

Catalog No. 50-002-60896 Supplier Medchemexpress LLC Supplier No. HY14184A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Macitentan (n-butyl analogue) is an n-butyl analogue of macitentan that acts as a dual endothelin ETA and ETB receptor antagonist. It is intended for research use in in vitro and preclinical pharmacology studies focused on endothelin signaling. Not for human or veterinary use.

  • Dual endothelin ETA and ETB receptor antagonist.
  • High purity (99.35%).
  • Molecular formula C20H21Br2N5O4S.
  • Soluble in DMSO; available as 10 mM solutions.
  • Available in small research pack sizes for screening.
  • Suitable for in vitro and preclinical pharmacology studies.
  • Provided as solid and solution formats for flexible use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.