Learn More
Vector Laboratories Maclura Pomifera Lectin (MPL), Unconjugated, 5mg
SDP
Catalog No. 502045678
Encompass
There appear to be two different chains in combinations giving rise to five isolectins. The carbohydrate specificity of either type of subunit appears to be the same, preferring α-linked N-acetylgalactosamine structures.Inhibiting Sugar: 500 mM galactose
Catalog No. 50-204-5678 Supplier Vector Laboratories Supplier No. L13405
There appear to be two different chains in combinations giving rise to five isolectins. The carbohydrate specificity of either type of subunit appears to be the same, preferring α-linked N-acetylgalactosamine structures.Inhibiting Sugar: 500 mM galactose