Learn More
Bioworld Maclura pomifera (Osage orange) Lectin (MPL/MPA) - Texas Red
Catalog No. 50199156
Encompass
Technical Specifications:Ligand Source:Maclura pomiferaMolecular Weight:44 kDaCarbohydrate Specificity:GalNAc, Galβ1,3GalNAcInhibiting/Eluting Sugar: GalactoseCarbohydrate for Elution:0.2M GalactoseBlood Group Specificity:A, B, O (-SA)Conjugate: Texas RedForm:LiquidIsoelectric (pI):4.8-5.3Storage: -20°CApplication note: N/A
Catalog No. 50-199-156 Supplier Bioworld Supplier No. 217610411
Technical Specifications:Ligand Source:Maclura pomiferaMolecular Weight:44 kDaCarbohydrate Specificity:GalNAc, Galβ1,3GalNAcInhibiting/Eluting Sugar: GalactoseCarbohydrate for Elution:0.2M GalactoseBlood Group Specificity:A, B, O (-SA)Conjugate: Texas RedForm:LiquidIsoelectric (pI):4.8-5.3Storage: -20°CApplication note: N/A