Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Macmillan Imidazolidinone Organocatalysts Kit I

Catalog No. 6745751KT Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-457-51KT 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 67-457-51KT Supplier Sigma Aldrich Supplier No. 6745751KT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

Specifications

CAS 32354-50-0
Quantity 500 mg
Synonym Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene][3-(2-pyridinyl) propylidene]ruthenium(II); Grubbs Catalyst(R) C598; [1,3-Dimesityl-2-imidazolidinylidene]dichloro[3-(2-pyridinyl)propylidene]ruthenium(II)
Molecular Weight (g/mol) 597.58 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.