Learn More
Macopharma Phlebotomy Bags
Catalog No. 23111002
Change view
Click to view available options
Includes:
16-gauge Needle
Luer Lock
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23-111-002 16-gauge Needle
23-111-003 Luer Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-111-002 Supplier Macopharma Supplier No. VSL7001PD
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Phlebotomy Bags
  • Increase patient safety and comfort during therapeutic phlebotomies
  • No vacuum required (unlike collection bottles), which reduces patient pain
  • Volume markings allow for accurate collections without a scale or mixer
  • Luer lock or 16-gauge needle configuration
  • Pull samples easily off of the collection; the product features a pre-connected vacuum tube adapter
  • Bags marked in 100mL increments allow for the precise collection of volume ordered by the physician
  • No risk of transfusing the blood; no outlet ports
  • Five-year shelf life (shelf life of standard blood bags is two years)
  • Individually wrapped bags allow for full shelf life
Order Info
  • 23-111-002 is a direct replacement for 23-001-000
  • 23-111-003 is a direct replacement for 23-001-001
  • 23-111-004 is a direct replacement for 23-001-002
Specifications
Product Type Phlebotomy Bag
Capacity (Metric) 600 mL
Includes 16-gauge Needle
No. per Case 20/Cs.