Learn More
Medchemexpress LLC HY-12903 5mg Medchemexpress, Macozinone CAS:1377239-83-2 Purity:>98%
SDP
Catalog No. 502081195
Encompass_Preferred
Medchemexpress, HY-12903 5mg Macozinone CAS:1377239-83-2 Macozinone (PBTZ169) is a bactericidal benzothiazinone and a potent DprE1 (decaprenylphosphoryl-β-d-ribose 2′-oxidase) inhibitor. Macozinone inhibits the essential flavoprotein DprE1 by forming a covalent bond with the active-site Cys387 residue. Macozinone has antituberculosis effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1195 Supplier Medchemexpress LLC Supplier No. HY129035MG
Medchemexpress, HY-12903 5mg Macozinone CAS:1377239-83-2 Macozinone (PBTZ169) is a bactericidal benzothiazinone and a potent DprE1 (decaprenylphosphoryl-β-d-ribose 2′-oxidase) inhibitor. Macozinone inhibits the essential flavoprotein DprE1 by forming a covalent bond with the active-site Cys387 residue. Macozinone has antituberculosis effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.