Learn More
Medchemexpress LLC Macranthoidin A | 140360-29-8 | 5 MG
SDP
Catalog No. 5000420323
Encompass_Preferred
Macranthoidin A is an orally active saponin from Flos Lonicerae. It offers protective effects against hepatic injury caused by Acetaminophen, Cd, and CCl4, and demonstrates noticeable depressant effects on ear croton oil swelling.
  • Orally active saponin from Flos Lonicerae.
  • Possesses protective effects on hepatic injury caused by Acetaminophen, Cd, and CCl4.
  • Exhibits conspicuous depressant effects on swelling of ear croton oil.
Catalog No. 50-004-20323 Supplier Medchemexpress LLC Supplier No. HYN21095MG
Macranthoidin A is an orally active saponin from Flos Lonicerae. It offers protective effects against hepatic injury caused by Acetaminophen, Cd, and CCl4, and demonstrates noticeable depressant effects on ear croton oil swelling.
  • Orally active saponin from Flos Lonicerae.
  • Possesses protective effects on hepatic injury caused by Acetaminophen, Cd, and CCl4.
  • Exhibits conspicuous depressant effects on swelling of ear croton oil.