Learn More
Medchemexpress LLC Macranthoidin A | 140360-29-8 | 99.4% | 1237.38 | C59H96O27 | 10 MG
SDP
Catalog No. 5000295620
Encompass_Preferred
Macranthoidin A is an orally active saponin isolated from Flos Lonicerae, supplied as an analytical standard for research use with reported hepatoprotective and anti-inflammatory activities.
  • Orally active saponin isolated from Flos Lonicerae.
  • Reported protective effects against acetaminophen-, cadmium-, and CCl4-induced liver injury.
  • Demonstrates depressant activity on ear croton oil swelling.
  • High purity suitable for analytical and pharmacological studies.
  • Available in small-mass packaging for research applications.
Catalog No. 50-002-95620 Supplier Medchemexpress LLC Supplier No. HYN210910MG
Macranthoidin A is an orally active saponin isolated from Flos Lonicerae, supplied as an analytical standard for research use with reported hepatoprotective and anti-inflammatory activities.
  • Orally active saponin isolated from Flos Lonicerae.
  • Reported protective effects against acetaminophen-, cadmium-, and CCl4-induced liver injury.
  • Demonstrates depressant activity on ear croton oil swelling.
  • High purity suitable for analytical and pharmacological studies.
  • Available in small-mass packaging for research applications.