Learn More
Cayman Chemical Macranthoidin B, Purity: ≥98%, CAS: 136849-88-2, 10 mg
SDP
Catalog No. 502990988
Encompass_Preferred
A triterpenoid saponin with anticancer activity; increases the production of ROS and induces apoptosis in HCT116 cells from 20-400 µM
Catalog No. 50-299-0988 Supplier Cayman Chemical Supplier No. 3153310 MG
A triterpenoid saponin with anticancer activity; increases the production of ROS and induces apoptosis in HCT116 cells from 20-400 µM