Learn More
Electron Microscopy Sciences Macro Cassette, White
SDP
Catalog No. 501899230
Encompass_Preferred
Made from Acetal-Polymer. Comes complete with a one piece integral lid . It is specially designed to hold large specimens during the embedding process, as well as in a storage cabinet. Inside measurements: 29 mm(L) x 25 mm(W) x 11 mm(D).
Color: White
Catalog No. 50-189-9230 Supplier Electron Microscopy Sciences Supplier No. 70077W
Made from Acetal-Polymer. Comes complete with a one piece integral lid . It is specially designed to hold large specimens during the embedding process, as well as in a storage cabinet. Inside measurements: 29 mm(L) x 25 mm(W) x 11 mm(D).
Color: White