Learn More
CELLTREAT Scientific Products 5mL Macro Centrifuge Tube, Snap-Pop, Assorted Color, Non-sterile
SDP
Catalog No. NC3377704
Encompass_Preferred
Offers the convenience of a micro centrifuge tube with the ability to process sample volumes up to 5mLTube diameter is the same as a standard 15mL conical bottom centrifuge tube enabling use with existing equipmentAll tubes have frosted marking spot and engraved graduationsSnap-Pop lids latch securely in place with a gentle pushCenter of the cap includes a thin “membrane” area designed for easy puncture and access by a syringe/needleColor tubes allow for quick sample identification, or to customize workflows
Catalog No. NC3377704 Supplier CELLTREAT Scientific Products Supplier No. 229445A
Offers the convenience of a micro centrifuge tube with the ability to process sample volumes up to 5mLTube diameter is the same as a standard 15mL conical bottom centrifuge tube enabling use with existing equipmentAll tubes have frosted marking spot and engraved graduationsSnap-Pop lids latch securely in place with a gentle pushCenter of the cap includes a thin “membrane” area designed for easy puncture and access by a syringe/needleColor tubes allow for quick sample identification, or to customize workflows