Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Macro Mania
SDPSCI_ED

Catalog No. S85018 Shop All LaMotte Company Products
Change view
Click to view available options
Format:
Game
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S85018 Game
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S85018 Supplier LaMotte™ Supplier No. 5942
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Cards (six decks), sorting sheets (6), instruction manual, and colorful Macro Mania classroom poster. Optional Expansion Kit (5943) includes: cards (three decks) and sorting sheets (3).

Classroom adventure introduces the use of stream macroinvertebrates to determine water quality.

  • Students learn about the influence of land use on water quality and the practice of macroinvertebrate sampling to measure water quality
  • Contains background information and step-by-step instructions for activities and extensions
  • Integrates math, public speaking, writing and art

Order Info

Class size: six teams. Expansion kit adds three more teams.

Specifications

Format Game
Product Type Macro Mania Adventure
Includes Cards (6 decks), sorting sheets (6), instruction manual and colorful Macro Mania classroom poster. Optional Expansion Kit (5943) includes: cards (3 decks) and sorting sheets (3)
For Use With (Application) Introduce the use of stream macroinvertebrates to determine Water quality. Lecture material sets the scene for the activity
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.