Learn More
Marketing International Macro-Prep Disposable Knives
Catalog No. 22284982
Change view
Click to view available options
Length (English) Blade:
8 in.
10 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Blade
22-284982 -
22-284980 10 in.
22-234682 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-284982 Supplier Marketing International Supplier No. 5014
No reprocessing with all-in-one disposable blade and handle
  • Longer-lasting, double-honed edge
  • Sheffield stainless-steel blades
  • Shipped 10 per pack
Specifications
Blade Material Stainless Steel
Product Type Macro Knife
Disposable Single-use