Learn More
Bio Rad Laboratories MACRO-PREP HIGH Q SUPPORT
Supplier:  Bio Rad Laboratories 1580040
Encompass
High Q Support; Bio-Rad; Macro-Prep; High capacity for biomolecules; Rigid polymer matrix; Macroporous beads that enhance access to ionic sites; High chemical, mechanical, and thermal stability; 25mL
Catalog No. 50-901-2470