Learn More
StatLab Macro-Preparation Grossing Knives
SDP
Catalog No. 23640016
Change view
Click to view available options
Length (English):
4 in.
6 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
23-640-016 4 in.
23-640-020 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-640-016 Supplier StatLab Supplier No. 5004
Add to Cart
Add to Cart
Built for dependable performance in routine pathology preparation tasks
  • Disposable knives intended for autopsy and gross examination workflows in histology and pathology laboratories, supporting precise tissue handling.
  • Integrated handle surface helps reduce slippage during use, contributing to safer and more controlled cutting.
  • Blade edge may be re-sharpened to help extend functional lifespan during laboratory procedures.
  • Packaged in boxes of ten to support efficient inventory management and daily lab operations.
Specifications
Length (English) 4 in.