Learn More
StatLab™ Macro-Preparation Knife
SDP
Catalog No. 22222167
Change view
Click to view available options
Length (English) Blade:
8 in.
10 in.
12 in.
Product Type:
Macro-autopsy Knife
Macro-preparation Brain Knife
Macro-preparation Lung Knife
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Blade Product Type
22-222-167 8 in. Macro-autopsy Knife
22-222-168 10 in. Macro-preparation Lung Knife
22-222-169 12 in. Macro-preparation Brain Knife
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-222-167 Supplier StatLab™ Supplier No. 5000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Features a disposable blade with an integral non-slip handle for autopsy and grossing.
  • May be re-sharpened for a longer lasting edge
Specifications
Product Type Macro-autopsy Knife
Length (English) Blade 8 in.
Disposable Single-use
Sterility Non-sterile