Learn More
Thomas Scientific 1.2ml Microtiter Cluster Tube, Nat NS, Bulk
SDP
Catalog No. NC9295705
Encompass_Preferred
1.2ml Macrotubes, Polypropylene, Bulk 1000/bag, 8.8 x 45mm plain tubes, autoclavable, sealable and freezable. Designed for use with 96 well macrotube plates.
Catalog No. NC9295705 Supplier Thomas Scientific Supplier No. 1148B58
1.2ml Macrotubes, Polypropylene, Bulk 1000/bag, 8.8 x 45mm plain tubes, autoclavable, sealable and freezable. Designed for use with 96 well macrotube plates.