Learn More
Medchemexpress LLC HY-111895A 10mg Medchemexpress, Macropa-NH2 (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY111895A10MG
Encompass_Preferred
Medchemexpress, HY-111895A 10mg Macropa-NH2 (HCl) CAS: Macropa-NH2 hydrochloride is the precursor of Macropa-NCS. Macropa-NCS is conjugated to trastuzumab as well as to the prostate-specific membrane antigen-targeting compound RPS-070 and is a promising therapeutic radionuclide applied in the treatment of soft-tissue metastases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8980