Learn More
Enzo Life Sciences Macrophage marker monoclonal antibody (HAM56) (0.5ml)
SDP
Supplier:  Enzo Life Sciences ENZC34935
Encompass_Preferred
Clone: HAM56. Isotype: IgM. Immunogen: Sonicated human alveolar macrophages. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Monkey. Specificity: Recognizes fixed macrophages such as "tingible macrophages", interdigitating macrophages of lymph nodes or tissue macrophages. Applications: IHC (PS). Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.1% sodium azide and 1% BSA.
Catalog No. 50-201-3372