Learn More
Promocell GmbH PromoCell Macrophage Base Medium XF 250 ml, with C-39857 attached to flask
Catalog No. NC0692442
Encompass
The Macrophage Base Medium XF is the user-customizable version of the Macrophage Generation Media XF and does not include cytokines. It comes without cytokines as a universally applicable MDM culture system featuring a fully user-customizable macrophage differentiation and activation process.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine macrophages.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:Basal Medium (250ml)SupplementMix (for 250ml)Note: Requires additional supplementation with M-CSF / GM-CSFMore details for  differentiation factor/activator combinations and recommended concentrations are available in our Application Note.
Catalog No. NC0692442 Supplier Promocell GmbH Supplier No. C28057
The Macrophage Base Medium XF is the user-customizable version of the Macrophage Generation Media XF and does not include cytokines. It comes without cytokines as a universally applicable MDM culture system featuring a fully user-customizable macrophage differentiation and activation process.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for murine macrophages.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.Components:Basal Medium (250ml)SupplementMix (for 250ml)Note: Requires additional supplementation with M-CSF / GM-CSFMore details for  differentiation factor/activator combinations and recommended concentrations are available in our Application Note.