Learn More
Sigma Aldrich Fine Chemicals Biosciences Macrophage Colony-Stimulat
Catalog No. 502904119
Encompass
Macrophage Colony-Stimulating factor (M-CSf) is a hematopoietic growth factor which modulates the proliferation differentiation and functional activation of monocytes. It is produced by fibroblasts endothelial cells and bone marrow stromal cells. MCSf stimulates the functions of mature monocytes and macrophages.
Catalog No. 50-290-4119 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M651810UG
Macrophage Colony-Stimulating factor (M-CSf) is a hematopoietic growth factor which modulates the proliferation differentiation and functional activation of monocytes. It is produced by fibroblasts endothelial cells and bone marrow stromal cells. MCSf stimulates the functions of mature monocytes and macrophages.