Learn More
Medchemexpress LLC Macrophage mannose receptor 1 Antibody (YA3531) | 50 UL
SDP
Catalog No. 5000442920
Encompass_Preferred
Macrophage mannose receptor 1 Antibody (YA3531) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Macrophage mannose receptor 1. It is suitable for a wide range of applications in research.
  • Rabbit-derived, non-conjugated, recombinant IgG monoclonal antibody
  • Targets macrophage mannose receptor 1
  • Affinity purified
  • Suitable for Western blot (WB), immunohistochemistry-paraffin (IHC-P), immunohistochemistry-frozen (IHC-F), immunocytochemistry/immunofluorescence (ICC/IF), immunofluorescence-tissue (IF-Tissue), multiplex immunohistochemical (mIHC), immunoprecipitation (IP), chromatin immunoprecipitation (ChIP), flow cytometry (FC), and enzyme linked immunosorbent assay (ELISA)
Catalog No. 50-004-42920 Supplier Medchemexpress LLC Supplier No. HYP8121450UL
Macrophage mannose receptor 1 Antibody (YA3531) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Macrophage mannose receptor 1. It is suitable for a wide range of applications in research.
  • Rabbit-derived, non-conjugated, recombinant IgG monoclonal antibody
  • Targets macrophage mannose receptor 1
  • Affinity purified
  • Suitable for Western blot (WB), immunohistochemistry-paraffin (IHC-P), immunohistochemistry-frozen (IHC-F), immunocytochemistry/immunofluorescence (ICC/IF), immunofluorescence-tissue (IF-Tissue), multiplex immunohistochemical (mIHC), immunoprecipitation (IP), chromatin immunoprecipitation (ChIP), flow cytometry (FC), and enzyme linked immunosorbent assay (ELISA)