Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Macrophage-SFM (1X)

Catalog No. 12065074
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-065-074 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-065-074 Supplier Gibco™ Supplier No. 12065074
  • $184.00
    $169.65 / Each of 1

    Save $14.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Macrophage-SFM (1X)

Macrophage-SFM is a serum-free medium designed for the culture of human peripheral blood monocytes and macrophages (1).

Contains L-glutamine, but no antibiotics.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Macrophages, Monocytes
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile-filtered
With Additives Glutamine
Quantity 500 mL
Shipping Condition Room Temperature
Endotoxin Level ≤1 EU/mL, ≤ 1 EU/mL
Included Antibiotics No antibiotics
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.