Learn More
Cole-Parmer Cole-Parmer Macropipettor, 2.0-10.0 mL
Supplier:  Cole-Parmer 0785940
Encompass_Preferred
Quickly adjust volume by turning the thumb knob until the yellow sight line corresponds to the desired setting on the volume window. Tighten a brass set screw, and your volume setting is locked in place. Polypropylene construction and durable design reduce friction wear and increase precision. A piston of PTFE ensures complete sealing in a polypropylene cylinder. O-rings are not exposed to friction wear. You can draw and deliver precise samples for the entire life of the piston and cylinder.
Catalog No. NC1695518