Learn More
Cytiva Macrosep Advance Centrifugal Devices With Supor Membrane - 0.45 µm, wildberry (24/pkg)
Supplier:  Cytiva MAPM45C67
Encompass
Quickly Concentrates Up To 20 mL of Biological Sample-Rapidly concentrates 20 mL sample volumes to 0.5 mL-Achieve 50X concentration and > 90% recovery in just minutes-Low protein-binding Omega™ membrane and polypropylene housing minimize losses due to non-specific binding-Versatile Omega membrane is available in a variety of MWCOs-Built-in deadstop prevents spinning to dryness-Color-coded and laser etched for easy identification This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9527