Learn More
AdipoGen Macrosphelide A (5 mg)
SDP
Supplier:  AdipoGen AGCN20152M005
Encompass_Preferred
Chemical. CAS: 172923-77-2. Formula: C16H22O8. MW: 342.4. Isolated from Paraconiothyrium sporulosum FO-5050. Antibiotic. Cell-cell adhesion inhibitor. Antimetastatic compound. Antifungal. Antimicrobial.
Catalog No. 50-205-3535