Learn More
AdipoGen Macrosporin (1 mg)
SDP
Supplier:  AdipoGen AGCN20497M001
Encompass_Preferred
Chemical. CAS: 22225-67-8. Formula: C16H12O5. MW: 284.3. Isolated from Stemphylium sp. Anthraquinone antibiotic. Moderate antibacterial agent. Photosensitizer (through ROS production) and leaf necrosis inducer. Antiproliferative. Moderate cytotoxic in selected cancer cells. Shown to inhibit several protein kinases, including Aurora B and FLT3 in low µM range. Mycotoxin. Useful as a reference in food analysis.
Catalog No. 50-205-3681