Learn More
Medchemexpress LLC Macupatide | 2923558-68-1 | >99.95% | 4984.60 | 10 MG
SDP
Catalog No. 5000422425
Encompass_Preferred
Macupatide is a glucose-dependent insulinotropic polypeptide (GIP) receptor agonist. It improves insulin secretion responses and insulin sensitivity, making it suitable for the research of type 2 diabetes.
  • Improves insulin secretion responses
  • Enhances insulin sensitivity
  • Produces larger insulin sensitivity improvement compared with selective GLP-1 RA Dulaglutide
  • Combination therapy with Dulaglutide has additive effects on insulin sensitivity
Catalog No. 50-004-22425 Supplier Medchemexpress LLC Supplier No. HYP1095910MG
Macupatide is a glucose-dependent insulinotropic polypeptide (GIP) receptor agonist. It improves insulin secretion responses and insulin sensitivity, making it suitable for the research of type 2 diabetes.
  • Improves insulin secretion responses
  • Enhances insulin sensitivity
  • Produces larger insulin sensitivity improvement compared with selective GLP-1 RA Dulaglutide
  • Combination therapy with Dulaglutide has additive effects on insulin sensitivity