Learn More
Medchemexpress LLC Macupatide acetate | 99.9% | 4984.60 | 1 MG
SDP
Catalog No. 5000424082
Encompass_Preferred
Macupatide acetate is a glucose-dependent insulinotropic polypeptide (GIP) receptor agonist that improves insulin secretion responses and insulin sensitivity. It can be used for the research of type 2 diabetes.
  • GIP receptor agonist
  • Improves insulin secretion responses
  • Enhances insulin sensitivity
  • Suitable for type 2 diabetes research
  • Produces larger insulin sensitivity improvement compared to selective GLP-1 RA Dulaglutide
  • Exhibits additive effects on insulin sensitivity when combined with other agents
Catalog No. 50-004-24082 Supplier Medchemexpress LLC Supplier No. HYP10959A1MG
Macupatide acetate is a glucose-dependent insulinotropic polypeptide (GIP) receptor agonist that improves insulin secretion responses and insulin sensitivity. It can be used for the research of type 2 diabetes.
  • GIP receptor agonist
  • Improves insulin secretion responses
  • Enhances insulin sensitivity
  • Suitable for type 2 diabetes research
  • Produces larger insulin sensitivity improvement compared to selective GLP-1 RA Dulaglutide
  • Exhibits additive effects on insulin sensitivity when combined with other agents