Learn More
Med Vet International MAD Nasal™ and MADgic Atomization Devices - MAD (w/ 3mL syringe & vial adapter), J1579A, Each
Catalog No. 5000585860
Encompass_Preferred
Medication Delivery Many water soluble medications can be safely administered intranasally. Micron particle size is 30-100 microns. Rapidly Effective The atomized particles are the optimal size for rapid absorption across mucosal membranes into the blood stream. Needle Free Provides painless, non-invasive rapid delivery. Controlled Administration Malleable tip atomizes in any position which helps with noncompliant patients. No sterile technique is required. Vial Adapter (Optional) The vial adapter has an end spike to penetrate rubber cap of medication vial. MAD Nasal™ and MADgic Atomization Devices - MAD (w/ 3ml syringe & vial adapter)
Catalog No. 50-005-85860 Supplier Med Vet International Supplier No. JOR1579A
Medication Delivery Many water soluble medications can be safely administered intranasally. Micron particle size is 30-100 microns. Rapidly Effective The atomized particles are the optimal size for rapid absorption across mucosal membranes into the blood stream. Needle Free Provides painless, non-invasive rapid delivery. Controlled Administration Malleable tip atomizes in any position which helps with noncompliant patients. No sterile technique is required. Vial Adapter (Optional) The vial adapter has an end spike to penetrate rubber cap of medication vial. MAD Nasal™ and MADgic Atomization Devices - MAD (w/ 3ml syringe & vial adapter)