Learn More
Aviva Systems Biology Corp MAD2L1 Antibody - middle region (AVARP04002_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP04002_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAD2L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAD2L1. Alternate names: HSMAD2, MAD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Mouse, Rat
Catalog No. 50-014-62532