Learn More
Aviva Systems Biology Corp MAD2L1BP Antibody - N-terminal region (ARP61356_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61356_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAD2L1BP. It was validated on Western Blot. . Alternate names: CMT2, KIAA0110, MGC11282, RP1-261G23.6ApplicationWB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-287-351