Learn More
MADA INC MadaCide-1 Disinfectant Cleaner | 4 x 1 Gal
Catalog No. NC1809270
Encompass_Preferred
MadaCide-1 is Mada Medical's disinfectant/cleaner/deodorizer that has no alcohol content. It is a general one-step, ready-to-use, hospital-level product. The product is effective bactericide, virucide, and fungicide. It is formulated to disinfect hard, non-porous, inanimate, environmental surfaces, equipment and non-critical instruments. Kills: Influenza A, HIV, Hepatitis A,B & C, TB, MRSA, VRE, and SARS.
Catalog No. NC1809270 Supplier MADA INC Supplier No. 7009
MadaCide-1 is Mada Medical's disinfectant/cleaner/deodorizer that has no alcohol content. It is a general one-step, ready-to-use, hospital-level product. The product is effective bactericide, virucide, and fungicide. It is formulated to disinfect hard, non-porous, inanimate, environmental surfaces, equipment and non-critical instruments. Kills: Influenza A, HIV, Hepatitis A,B & C, TB, MRSA, VRE, and SARS.