Learn More
MADA MEDICAL PRODUCTS INC MadaCide-FD, 4x1Gallon
Catalog No. NC9288230
Encompass_Preferred
MadaCide-FD is a hospital-level Disinfectant/Cleaner/Deodorizer that is designed specifically for the infection control needs of healthcare. Efficacy tests have demonstrated that this product is an effective bactericide, virucide, germicide, and fungicide. It is formulated to disinfect hard, non-porous, inanimate, environmental surfaces, including chrome, glass, painted surfaces, plastics and stainless steel. It is ideal for use with most medical surfaces such as operating rooms, respiratory therapy equipment and medical instruments. Unique anti-corrosion formulation will not damage lenses, cement, plastics, rubber, steel, aluminum, brass or other metal when used as directed. Kills Influenza A, HIV, Hepatitis B & C, TB, MRSA, VRE, and SARS.
This item is not returnable. View return policy
Catalog No. NC9288230 Supplier MADA MEDICAL PRODUCTS INC Supplier No. 7021
MadaCide-FD is a hospital-level Disinfectant/Cleaner/Deodorizer that is designed specifically for the infection control needs of healthcare. Efficacy tests have demonstrated that this product is an effective bactericide, virucide, germicide, and fungicide. It is formulated to disinfect hard, non-porous, inanimate, environmental surfaces, including chrome, glass, painted surfaces, plastics and stainless steel. It is ideal for use with most medical surfaces such as operating rooms, respiratory therapy equipment and medical instruments. Unique anti-corrosion formulation will not damage lenses, cement, plastics, rubber, steel, aluminum, brass or other metal when used as directed. Kills Influenza A, HIV, Hepatitis B & C, TB, MRSA, VRE, and SARS.