Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology MAdCAM-1 Monoclonal Antibody, Rabbit IgG, Human Reactivity, 100 μl
SDP

Catalog No. 502729967
Encompass

This rabbit monoclonal antibody is designed to detect total MAdCAM-1 protein in human samples. The antibody recognizes transfected levels of total MAdCAM-1 by western blot and endogenous levels of total MAdCAM-1 by immunohistochemistry (paraffin). Produced via immunization with a synthetic peptide corresponding to residues near the carboxy terminus of the human protein, it exhibits an observed molecular weight of 60–85 kDa, which deviates from the predicted 40 kDa due to glycosylation.

  • Host species: Rabbit
  • Reactivity: Human
  • Applications: Western blotting (1:1,000 dilution) and immunohistochemistry (paraffin) (1:100 to 1:400 dilution)
  • Format: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Storage: Store at −20°C; do not aliquot

Catalog No. 50-272-9967 Supplier Cell Signaling Technology Supplier No. 93757S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit monoclonal antibody is designed to detect total MAdCAM-1 protein in human samples. The antibody recognizes transfected levels of total MAdCAM-1 by western blot and endogenous levels of total MAdCAM-1 by immunohistochemistry (paraffin). Produced via immunization with a synthetic peptide corresponding to residues near the carboxy terminus of the human protein, it exhibits an observed molecular weight of 60–85 kDa, which deviates from the predicted 40 kDa due to glycosylation.

  • Host species: Rabbit
  • Reactivity: Human
  • Applications: Western blotting (1:1,000 dilution) and immunohistochemistry (paraffin) (1:100 to 1:400 dilution)
  • Format: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Storage: Store at −20°C; do not aliquot

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.