Learn More
MSC Made in USA Ball Twine, Cotton, 8 Ply, 24 lb Breaking Strength
Catalog No. 5000528221
Encompass
This ball twine is made from natural cotton and features an 8 ply construction. It offers a 24 lb breaking strength and comes with 2,469 feet per pound, suitable for various general-purpose applications.
  • Twine ball design
  • Cotton material
  • Natural color
  • 8 ply construction
  • 24 lb breaking strength
  • 2,469 feet per pound
  • Manufactured in the United States
Catalog No. 50-005-28221 Supplier MSC Supplier No. 45890670
This ball twine is made from natural cotton and features an 8 ply construction. It offers a 24 lb breaking strength and comes with 2,469 feet per pound, suitable for various general-purpose applications.
  • Twine ball design
  • Cotton material
  • Natural color
  • 8 ply construction
  • 24 lb breaking strength
  • 2,469 feet per pound
  • Manufactured in the United States