Learn More
MSC Made in USA Hardness Calibration Test Block, Rockwell B Scale, 40 HRB
Catalog No. 5000534210
Encompass
This product is a hardness calibration test block designed for use with the Rockwell B scale. It is specifically suited for low hardness range applications, providing a maximum hardness of 40 HRB.
  • Designed for Rockwell B scale hardness testing.
  • Features a maximum hardness of 40 HRB.
  • Suitable for low hardness range applications.
  • Manufactured in the USA.
Catalog No. 50-005-34210 Supplier MSC Supplier No. 00444745
This product is a hardness calibration test block designed for use with the Rockwell B scale. It is specifically suited for low hardness range applications, providing a maximum hardness of 40 HRB.
  • Designed for Rockwell B scale hardness testing.
  • Features a maximum hardness of 40 HRB.
  • Suitable for low hardness range applications.
  • Manufactured in the USA.