Learn More
MSC Made in USA Sash Chain, 0.035 in Diameter, Steel, Copper Dipped, 500 ft
Catalog No. 5000515554
Encompass
This 0.035 in diameter steel sash chain, manufactured in the USA, features a copper dipped finish and is designed for applications requiring a load limit of 105 Lb. It is supplied in a total coil length of 500 ft.
  • Steel material ensures durability.
  • Copper dipped finish provides corrosion resistance.
  • 0.035 in diameter.
  • Work load limit of 106 Lb.
  • Total coil length of 500 ft.
  • Trade size #35.
  • Chain grade 30.
  • Inside length of 0.5700 decimal "
  • 21.2 links per foot.
Catalog No. 50-005-15554 Supplier MSC Supplier No. 67777565
This 0.035 in diameter steel sash chain, manufactured in the USA, features a copper dipped finish and is designed for applications requiring a load limit of 105 Lb. It is supplied in a total coil length of 500 ft.
  • Steel material ensures durability.
  • Copper dipped finish provides corrosion resistance.
  • 0.035 in diameter.
  • Work load limit of 106 Lb.
  • Total coil length of 500 ft.
  • Trade size #35.
  • Chain grade 30.
  • Inside length of 0.5700 decimal "
  • 21.2 links per foot.