Learn More
Cayman Chemical MadecasIc AcId 100mg
SDP
Catalog No. 502951718
Encompass_Preferred
A triterpenoid with diverse biological activities; inhibits LPS-induced production of NO, PGE2, TNF-α, IL-1β, and IL-6 in RAW 264.7 macrophages at 150 UG/ml; decreases plasma levels of fibrinogenin and triglycerides, as well as heart and kidney levels of ROS, in a mouse model of streptozotocin-induced diabetes at (0.05 and 0.1% in the diet; decreases tumor growth in a CT26 murine colon cancer model
Catalog No. 50-295-1718 Supplier Cayman Chemical Supplier No. 11854100 MG
A triterpenoid with diverse biological activities; inhibits LPS-induced production of NO, PGE2, TNF-α, IL-1β, and IL-6 in RAW 264.7 macrophages at 150 UG/ml; decreases plasma levels of fibrinogenin and triglycerides, as well as heart and kidney levels of ROS, in a mouse model of streptozotocin-induced diabetes at (0.05 and 0.1% in the diet; decreases tumor growth in a CT26 murine colon cancer model