Learn More
Cayman Chemical MadecasosIde 10mg
SDP
Catalog No. 502955574
Encompass_Preferred
A natural triterpenoid with a wide range of reported anti-inflammatory, wound healing, and antioxidant activities; suppresses LPS-induced TNF-α production via inhibition of ERK, p38, and NF-κB activity
Catalog No. 50-295-5574 Supplier Cayman Chemical Supplier No. 1185510 MG
A natural triterpenoid with a wide range of reported anti-inflammatory, wound healing, and antioxidant activities; suppresses LPS-induced TNF-α production via inhibition of ERK, p38, and NF-κB activity