Learn More
Cayman Chemical Madecassoside, Purity: ≥98%, CAS: 34540-22-2, 50 mg
SDP
Catalog No. 502955781
Encompass_Preferred
A natural triterpenoid with a wide range of reported anti-inflammatory, wound healing, and antioxidant activities; suppresses LPS-induced TNF-α production via inhibition of ERK, p38, and NF-κB activity
Catalog No. 50-295-5781 Supplier Cayman Chemical Supplier No. 1185550 MG
A natural triterpenoid with a wide range of reported anti-inflammatory, wound healing, and antioxidant activities; suppresses LPS-induced TNF-α production via inhibition of ERK, p38, and NF-κB activity